Methyl-4-acetylbenzoate 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

424770010 424770050
ACRO424770010EA 360 DKK
ACRO424770010 ACRO424770050
Methyl-4-acetylbenzoate 99+%
Methyl-4-acetylbenzoate
Formel: C₁₀H₁₀O₃
Smeltepunkt: 93…95 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00216474
CAS-nummer: 3609-53-8

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 93 to 95 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR