(±)-Naringenin ≥93.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin
N0072-25G
N0072-5G
TCIAN0072-25GEA
1320
DKK
TCIAN0072-25G
TCIAN0072-5G
(±)-Naringenin ≥93.0% (by HPLC, titration analysis)
(±)-Naringenin
Formel:
C₁₅H₁₂O₅ |
MDL Number:
MFCD00006844 CAS-nummer: 67604-48-2 EINECS: 266-769-1 Merck Index: 14,06424 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Purity (Neutralization titration) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...