Hesperetin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin

B20528.03 B20528.06 B20528.14
B20528.03EA 321 DKK
B20528.03 B20528.06 B20528.14
Hesperetin 97%
Hesperetin

Purity: 97% CAS No.: 520-33-2 Molecular Formula: C16H14O6 Formula Weight: 302.30 Melting Point: 227-232° Beilstein Reference: 309850 Merck Reference: 14,4670 MDL No.: MFCD00075646

Formel: C₁₆H₁₄O₆
Molekylvægt: 302.30 g/mol
Smeltepunkt: 227…232 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075646
CAS-nummer: 520-33-2
EINECS: 208-290-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR