2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A1356-5G A1356-BULK
TCIAA1356-5GEA 386 DKK
TCIAA1356-5G TCIAA1356-BULK
2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-4',5'-methylenedioxyacetophenone
Formel: C₉H₉NO₃
MDL Number: MFCD00075855
CAS-nummer: 28657-75-2
EINECS: 249-131-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd1d82fcd3d047ae4548620dc710ffefa7445799&vid=cd1d82fcd3d047ae4548620dc710ffefa7445799&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR