Anisomycin
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
13640.01
13640.02
SERA13640.01EA
0
DKK
SERA13640.01
SERA13640.02
Anisomycin
Anisomycin
Formel:
C₁₄H₁₉NO₄ Molekylvægt: 265.31 g/mol |
MDL Number:
MFCD00077650 CAS-nummer: 22862-76-6 EINECS: 245-269-7 UN nummer: 3462 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...