1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1770-25G T1770-5G
TCIAT1770-25GEA 2270 DKK
TCIAT1770-25G TCIAT1770-5G
1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97.0% (by GC)
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Formel: C₁₄H₁₉F₁₃O₃Si
Flammepunkt: 97°C
MDL Number: MFCD00042333
CAS-nummer: 51851-37-7
EINECS: 257-473-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aff85c715a0eb10abcc887b0ec0ff7dc7d2781f&vid=3aff85c715a0eb10abcc887b0ec0ff7dc7d2781f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR