Allantoin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(2,5-Dioxoimidazolidin-4-yl)urea

A15571.30 A15571.0B A15571.0I
A15571.30EA 663 DKK
A15571.30 A15571.0B A15571.0I
Allantoin ≥98%
Allantoin

Purity: 98% CAS No.: 97-59-6 Molecular Formula: C4H6N4O3 Formula Weight: 158.12 Melting Point: ca 230° dec. Beilstein Reference: 102364 Merck Reference: 14,258 MDL No.: MFCD00005260

Formel: C₄H₆N₄O₃
Smeltepunkt: ∼ 230 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005260
CAS-nummer: 97-59-6
EINECS: 202-592-8
Merck Index: 14,258

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR