2-Hydroxy-5-methylbenzophenone ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H1008-25G H1008-5G
TCIAH1008-25GEA 1650 DKK
TCIAH1008-25G TCIAH1008-5G
2-Hydroxy-5-methylbenzophenone ≥97.0% (by GC)
2-Hydroxy-5-methylbenzophenone
Formel: C₁₄H₁₂O₂
MDL Number: MFCD00002379
CAS-nummer: 1470-57-1
EINECS: 216-002-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=250af38f0d8e2d7f1c3de55d754b9b643f0b156f&vid=250af38f0d8e2d7f1c3de55d754b9b643f0b156f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 81.0 to 85.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR