Ionomycin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

J62448.MX J62448.X0
J62448.MXEA 1020 DKK
J62448.MX J62448.X0
Ionomycin ≥98%
Ionomycin
Formel: C₄₁H₇₂O₉
Opbevaringstemperatur: Fryser
MDL Number: MFCD06798385
CAS-nummer: 56092-81-0
UN nummer: 1993
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR