Bromotrispyrrolidinophosphonium hexafluorophosphate
Leverandør: Novabiochem (Part of Merck)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
8.51010.0100 8.51010.0025
01-62-0017-100EA 12540 DKK
01-62-0017-100 01-62-0017-25
Bromotrispyrrolidinophosphonium hexafluorophosphate
Bromotrispyrrolidinophosphonium hexafluorophosphate
Formel: C₁₂H₂₄BrF₆N₃P₂
Molekylvægt: 466.2 g/mol
MDL Number: MFCD00077412
CAS-nummer: 132705-51-2