Lansoprazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

L0233-1G L0233-5G
TCIAL0233-1GEA 550 DKK
TCIAL0233-1G TCIAL0233-5G
Lansoprazole ≥98.0% (by HPLC, titration analysis)
Lansoprazole
Formel: C₁₆H₁₄F₃N₃O₂S
Molekylvægt: 369.36 g/mol
Smeltepunkt: 170°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00866873
CAS-nummer: 103577-45-3
Merck Index: 14,05362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d91e0a26f6bf6f0fbb0582cb7b38972ea4eec9f&vid=4d91e0a26f6bf6f0fbb0582cb7b38972ea4eec9f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR