Folic acid

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

T912.1 T912.2
ROTHT912.1EA 399.33 DKK
ROTHT912.1 ROTHT912.2
Folic acid
Folic acid
Formel: C₁₉H₁₉N₇O₆
Molekylvægt: 441.41 g/mol
MDL Number: MFCD00079305
CAS-nummer: 59-30-3
EINECS: 200-419-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR