Hesperidin ≥90.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
H0049-25G
H0049-500G
H0049-100G
TCIAH0049-25GEA
279
DKK
TCIAH0049-25G
TCIAH0049-500G
TCIAH0049-100G
Hesperidin ≥90.0% (by HPLC, titration analysis)
Hesperidin
Formel:
C₂₈H₃₄O₁₅ |
MDL Number:
MFCD00075663 CAS-nummer: 520-26-3 EINECS: 208-288-1 Merck Index: 14,04671 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Purity (Neutralization titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...