Carbamazepine 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
344360050
344360250
344360010
ACRO344360050EA
847
DKK
ACRO344360050
ACRO344360250
ACRO344360010
Carbamazepine 98%
Carbamazepine
Formel:
C₁₅H₁₂N₂O Smeltepunkt: 189…192 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00005073 CAS-nummer: 298-46-4 EINECS: 206-062-7 |
Specifikationer Testresultater
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 189 to 193 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...