[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), [1,1’-Bis(diphenylphosphino)ferrocene]dichloronickel(II)
B2226-1G
B2226-5G
TCIAB2226-1GEA
427
DKK
TCIAB2226-1G
TCIAB2226-5G
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥97.0% (by titrimetric analysis)
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride
Formel:
C₃₄H₂₈Cl₂FeNiP₂ Molekylvægt: 683.98 g/mol |
MDL Number:
MFCD00270284 CAS-nummer: 67292-34-6 |
|
Specifikationer Testresultater
Appearance | Green to Dark green powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...