4-Methoxyphenyl chloroformate 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
314570050
314570250
ACRO314570050EA
593
DKK
ACRO314570050
ACRO314570250
4-Methoxyphenyl chloroformate 98%
4-Methoxyphenyl chloroformate
Formel:
C₇H₄BrClO₂ Kogepunkt: 49 °C (,2 mmHg) Massefylde: 1,25 g/cm³ Flammepunkt: >110 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00013258 CAS-nummer: 7693-41-6 UN nummer: 3277 ADR: 6.1,II |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5220 to 1.5260 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...