N-Bromoacetamide ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B0530-25G B0530-5G
TCIAB0530-25GEA 1400 DKK
TCIAB0530-25G TCIAB0530-5G
N-Bromoacetamide ≥97.0% (by titrimetric analysis)
N-Bromoacetamide
Formel: C₂H₄BrNO
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00037097
CAS-nummer: 79-15-2
EINECS: 201-181-0
UN nummer: 1759
ADR: 8,III
Merck Index: 14,01398
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=401e461b87b65b402ac71623bab3863dd1b2b2e4&vid=401e461b87b65b402ac71623bab3863dd1b2b2e4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 105.0 to 109.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR