(+)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (1S,2R,5S)-2-Isopropyl-5-methylcyclohexyl carbonochloridate, (1S)-(+)-Menthyl chloroformate

M1221-25ML M1221-5ML
TCIAM1221-25MLEA 1260 DKK
TCIAM1221-25ML TCIAM1221-5ML
(+)-Menthyl chloroformate ≥97.0% (by titrimetric analysis)
(+)-Menthyl chloroformate
Formel: C₁₁H₁₉ClO₂
Molekylvægt: 218.720 g/mol
Flammepunkt: 70°C
MDL Number: MFCD00134483
CAS-nummer: 7635-54-3
UN nummer: 3277
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26ac42bcc168643601725c5969a787713e41a1fa&vid=26ac42bcc168643601725c5969a787713e41a1fa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 97.0 %
Specific rotation [a]20/D +70.0 to +80.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR