Erlenmeyer flasks, with PP screw cap, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FDE444 FDE446 FDE448 FDE450
214-0342EA 263 DKK
214-0342 214-0343 214-0344 214-0345
Erlenmeyer flasks, with PP screw cap, Azlon®
Kolber Erlenmeyer-kolber
PMP, transparent.
  • Good chemical resistance
  • Heat resistant up to 150 °C (up to 170 °C for short periods)
  • Flask and cap can be autoclaved at 121 °C
With red, easy to read graduations. Wide mouth with screw cap GL 40 or GL 52, can also be closed using standard ground PP stoppers.