Specimen containers, with captive hinged cap, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BGG306 BGG308 BGG312
216-1348EA 215 DKK
216-1348 216-1349 216-1351
Specimen containers, with captive hinged cap, Azlon®
Prøvecontainere Allround kasser
LDPE, translucent.
  • Good chemical resistance
  • Lightweight and unbreakable in normal use
  • Can withstand temperatures up to 80 °C
Containers with captive hinged cap suitable for many applications.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904