Cut resistant gloves, uvex C500 wet
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6049210 6049209 6049208 6049207 6049211
112-4493EA 1600 DKK
112-4493 112-4492 112-4491 112-4490 112-4428
Cut resistant gloves, uvex C500 wet
Handsker
These bamboo-rayon/Dyneema®/glass/polyamide gloves feature a HPE coating on palm and fingertips. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort.
  • Resistant to contact heat up to 100 °C
  • Oil and grease resistant
  • Outstanding wearing comfort and tactile feel with climazone technology
  • Very good grip in damp and wet environments
  • Highest cut resistance level 5
  • Silicone-free according to imprint test
Typical applications include: automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing, metal industry, assembly works, paper industry, shipping/logistics, transportation works.
Certificeringer: EN 388:2016 (4X42C)
OEKO-TEX® Standard 100