Tubing connectors, straight
Leverandør: SEMADENI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0504 0508 0509 0507 0506 0505
229-7483EA 26.2 DKK
229-7483 229-7484 229-7487 SEMA0507 SEMA0506 SEMA0505
Tubing connectors, straight
Fittings og stik
Connectors, made of white PP, with tapered ends, for connecting tubing with the same diameter.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable