Magnetic stirring bars, cylindrical
Leverandør: SEMADENI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2209 0244 0257 0258 0259 0320
SEMA2209EA 220 DKK
SEMA2209 SEMA0244 SEMA0257 SEMA0258 SEMA0259 SEMA0320
Magnetic stirring bars, cylindrical
Omrørere Omrøremagneter
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
  • Chemical resistance
  • Smooth surface and round ends
  • Temperature resistance upto 275 °C