Funnels, with short stem, PP
Leverandør: SEMADENI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0219 0221
221-1150EA 132 DKK
221-1150 SEMA0221
Funnels, with short stem, PP
Tragte Filtreringstragte Filtertragte
Funnels, made of PP, translucent, with slim stem.
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable