Cervical brushes, Cervex-Brush®
Leverandør: Rovers
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
380100324 380101010 380100331
720-2316EA 2410 DKK
720-2316 720-2319 720-2317
Cervical brushes, Cervex-Brush®
Børster
The reliability of smears obtained by the Cervex-Brush® reduces the need for repeat testing. Cell collection with the Cervex-Brush® provides a clear and accurate sample which leads to early detection of cervical cancer. Use of the Cervex-Brush® in preventative investigation reduces the need for more drastic measures to be taken. This represents enormous savings potential toward national health care costs.
  • Softness of the material ensures that no internal injury occurs eliminating blood admixutre and unsatisfactory smears as a result
  • Comfortable investigation method for both patient and doctor, which above all delivers a visibly good result
  • Can be used for liquid-based as well as conventional cytology
  • Hydrophobic material of the brush facilitates the release of the cell material into the fluid
The top portion uses a soft flexible brush to obtain cell samples. The brush consists of 57 semi-circular plastic bristles of various lengths. The shape is such that the top edges of the brush follow the contours of the cervix. The longer middle bristles reach deep into the endocervical canal. The shorter bristles touch both the ectocervical area and transformation zone at the same time.
Cervex-Brush® Combi: The convenient Cervex-Brush® Combi provides doctors and nurses the option of a high quality Pap smear collection with a single device. It enables simultaneous collection of cells of the ectocervix, endocervix and transformation-zone in a single sample without bleeding or pain. The Cervex-Brush® Combi is an improvement over the regular Cervex-Brush® as it collects 2-3 times more endocervical cells with only 2 clockwise rotations on the cervical os.
The Cervex-Brush® Combi can be used for HPV testing, conventional cytology and liquid-based cytology. The hydrophobic material of the brush facilitates the release of the cell material into the fluid.