Pipette Boxes, with Lid
Leverandør: BELLCO GLASS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1217-00409 1217-11109 1217-11116 1217-22216 1217-22218 1217-90218 1217-11106
BELC1217-00409EA 727.78 DKK
BELC1217-00409 BELC1217-11109 BELC1217-11116 BELC1217-22216 BELC1217-22218 BELC1217-90218 BELC1217-11106
Pipette Boxes, with Lid
Bokse Beholdere til Pipettesterilisering
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
  • Stackable
  • Silicone pad on inside of top and bottom protect pipette tips
  • Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.