VWR® Q Path® MicroStar III, Cassettes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2087EA 1400 DKK
720-2087 720-2088 720-2089 720-2081 720-2082 720-2083 720-2084 720-2086 720-2085
VWR® Q Path® MicroStar III, Cassettes
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Disposable lid, supplied in dispenser for 500 cassettes. Packed in three boxes of 500 cassettes.
  • Small opening for the filtration of fluids
  • Mounted lid with hinges
  • Writing area 45° angle suitable for all current printers