VWR® Q Path® MicroStar II, Cassettes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2073EA 1550 DKK
720-2073 720-2074 720-2075 720-2076 720-2077 720-2078 720-2079 720-2080 720-2072
VWR® Q Path® MicroStar II, Cassettes
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with separate disposable lid, supplied in distributing box with 2×1000 cassettes and 1×2000 lids. This cassette has been specially designed for cassette burners.
  • Easy to use cassette with small opening
  • Hinge and separate lid
  • With 45° writing angle
  • Cassettes with ridges, which makes it easier to be opened after impregnation