VWR® Q Path® MicroStar II, Cassettes, in sleeve
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2042EA 4890 DKK
720-2042 720-2043 720-2044 720-2045 720-2046 720-2047 720-2048 720-2049 720-2041
VWR® Q Path® MicroStar II, Cassettes, in sleeve
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes in pre-filled sleeves for Thermo and RA Lamb cassette printers. 50 dispensers with 80 cassettes and 2×2000 lids.
  • Small opening for the filtration of fluids
  • Hinge and separate lid
  • With 45° writing angle