VWR® Q Path® MacroStar V, Cassettes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2219EA 1160 DKK
720-2219 720-2220 720-2215 720-2221 720-2222 720-2223 720-2216 720-2218 720-2217
VWR® Q Path® MacroStar V, Cassettes
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded disposable lid, supplied in dispensers for 500 cassettes. Packed in three boxes of 500 cassettes.
  • Easy to use cassette
  • 45° writing area at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation