VWR® Q Path® MacroStar III, Cassettes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2238EA 1240 DKK
720-2238 720-2239 720-2240 720-2233 720-2234 720-2235 720-2236 720-2237 720-2241
VWR® Q Path® MacroStar III, Cassettes
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Disposable lid, supplied in dispenser for 500 cassettes. Packed in three boxes of 500 cassettes.
  • Large opening for the filtration of fluids
  • Mounted lid with hinges
  • Writing area 45° angle suitable for all current printers