VWR® Q Path® MacroStar I, Cassettes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2247EA 1790 DKK
720-2247 720-2248 720-2249 720-2250 720-2246 720-2244 720-2245 720-2242 720-2243
VWR® Q Path® MacroStar I, Cassettes
Histologikassetter Indstøbningskassetter, Biopsiprøver
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Accepts standard stainless steel lids. Box of 4000 cassettes.
  • Large opening for the filtration of fluids
  • With 45° writing angle
  • Without lid
  • With 34 holes