Solid phase extraction cartridges, HyperSep™ Florisil
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60108-735 60108-431 60108-405 60108-402 60108-403 60108-404 60108-500 60108-736 60108-737
549-0711EA 1440 DKK
549-0711 549-0709 549-0708 549-0705 549-0706 549-0707 549-0710 549-0712 549-0713
Solid phase extraction cartridges, HyperSep™ Florisil
Kromatografikolonner
Ideal for the isolation of polar compounds from non polar matrices.
  • Magnesia-loaded silica gel offers rapid flow rates for large sample volumes
  • Extremely polar in nature, especially suited to the separation of chlorinated pesticides