VWR® Q Path®, Biopsy Pads
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2254EA 289 DKK
720-2254 720-2255
VWR® Q Path®, Biopsy Pads
Biopsi skum indsats
Polyester urethane foam. To hold small biopsies in place and prevent them from being lost. Enables fluids to circulate freely during processing.
  • Individually cut
  • Ready to use
  • Solvent resistant
Packed in bags of 500 pieces.