VWR®, Filter Cards for Cytology Centrifuges
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
720-2059EA 536 DKK
720-2059 720-2060 720-2061
VWR®, Filter Cards for Cytology Centrifuges
Filtre Centrifugefiltre
Cyto strips made of filter board for cytocentrifuges. Absorption of excess liquids from the staining of samples in cytologic analysis. Can be used in Shandon and Sakura Cyto-Tek® cytocentrifuges.
  • For single and multiple chambers