Labeling tapes, warning tapes
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
255313 255297 255312
BRDY255313EA 399 DKK
BRDY255313 116-0231 BRDY255312
Labeling tapes, warning tapes
Tape
Self-adhesive warning film for marking pillars, pylons, the edges of walls, gate arches and lifts.
  • Extremely durable
  • Waterproof, oil, alkali and acid resistant
  • Different colours available
Length: 33,0 m.