HPLC columns, Chromolith® CN
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.52046.0001 1.52047.0001 1.52048.0001 1.52050.0001
1.52046.0001EA 4580 DKK
1.52046.0001 1.52047.0001 1.52048.0001 1.52050.0001
HPLC columns, Chromolith® CN
Kromatografikolonner
Cyano columns are generally more polar than traditional alkyl silica columns. The functional groups are highly ordered, reducing steric hindrance for the solute. The modification also allows cation exchange activity, which is higher at neutral pH than in acidic conditions. Chromolith® CN columns are suitable for the separation of alkaloids, oils, flavonoids, glycols, phenols, phthalates, steroids and sulfonamides.