Spigots
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0525-1000 0525-5000 0525-3000 0525-7000
216-2854EA 464 DKK
216-2854 216-0941 216-0940 216-0942
Spigots
Stophaner
Media-contacting parts PP, PE and PTFE.
  • Tight closing for complete seal
  • Easy to use
  • With connecting cap nut
The spigots fit all containers with R¾" connection threads. Screw thread adapters facilitate connection of spigots to nearly all containers. Pressure resistant up to 2 bar at 20 °C (without discharge bracket and grommet). With discharge angle and rotatable spout preventing drips. With tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm.
Not suitable for solidifying substances.