One-piece fingertight fittings
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AL32235 AL32236 AL32347
VARIAL32235EA 0 DKK
VARIAL32235 VARIAL32236 VARIAL32347
One-piece fingertight fittings
Fittings og stik
PEEK fittings resists pressure up to 5000 psi and temperature of 100 °C.