Production and laboratory chairs, with permanent contact backrest, Neon 2
Leverandør: bimos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9563-MG01-3278 9563-SP02-3280 9563-2000-3278 9563-2000-3279 9563-2000-3280 9563-2001-3278 9563-2001-3280 9563-MG01-3279 9563-MG01-3280 9563-MG02-3278 9563-MG02-3279 9563-MG02-3280 9563-SP01-3278 9563-SP01-3279 9563-SP01-3280 9563-SP02-3278 9563-2001-3279 9563-SP02-3279 9563-2002-3278 9563-2002-3279 9563-2002-3280 9563-MG11-3279 9563-MG11-3280 9563-MG11-3278 9563-SP11-3278 9563-SP11-3279 9563-SP11-3280 9563-6801-3280 9563-6801-3279 9563-6801-3278 9563-6802-3278 9563-6802-3279 9563-6802-3280 9563-6811-3278 9563-6811-3279 9563-6811-3280 9563-2000-3277 9563-6801-3277 9563-MG01-3277 9563E-98113280 9563E-MG013280 9563E-MG013278 9563E20003278 9563-SP01-3277 9563MG01327852 9563E-98013278 9563-SP02-3277 9563E20003279 9563EMG013279 9563E20003277 9563E20003280
132-0913EA 6910 DKK
132-0913 139-0657 132-0910 132-0911 132-0912 139-0631 139-0633 132-0914 132-0915 139-0643 139-0644 139-0645 139-0649 139-0653 139-0654 139-0655 139-0632 139-0656 139-0634 139-0635 139-0636 139-0647 139-0648 139-0646 139-0658 139-0659 139-0660 139-0437 132-0917 132-0916 139-0637 139-0638 139-0639 139-0640 139-0641 139-0642 132-0979 132-0981 132-0980 BIMO9563E-98113280 132-1416 132-1415 132-1414 BIMO9563-SP01-3277 132-1417 BIMO9563E-98013278 BIMO9563-SP02-3277 BIMO9563E20003279 BIMO9563EMG013279 BIMO9563E20003277 BIMO9563E20003280
Production and laboratory chairs, with permanent contact backrest, Neon 2
Stole
Neon supports the forward leaning sitting position that is often necessary in lab work. Its modern design matches the high-tech laboratory environment perfectly and has all the characteristics you would expect of a good laboratory chair. It is seamless, easy to clean, washable, and resistant to disinfectants making Neon the ideal solution for the laboratory when a really comfortable ergonomic chair is needed.
  • Castors
  • Permanent contact backrest
  • Ergonomic package with height, seat inclination, seat depth and backrest height adjustment and, weight regulation
  • Robust design for a long service life
  • Very few composite materials
  • Easy change of upholstery
  • Flex strip available in three colours
  • Five-star base made of aluminium; powder coated black
  • Anti-shock lock
  • Certified sustainable production
Permanent contact backrest: It follows the user's movements and ensures that the back always remains fully supported. Weight regulation ensures that the balance between the resistance of the backrest and the body weight is always steady. The permanent contact backrest is ideal for work where people have to lean forward for a long time, with only small distance between the eyes and workpiece (e.g. working at close range in a laboratory).
Certificeringer: Blue Angel Eco Label
Non toxic
Complies with DIN 68 877 standards
Conforms to quality management system DIN EN ISO 9001
Complies with regulation EWG No. 1836/93 and Environmental Management System DIN EN ISO 14001