PIG® encapsulating and neutralizing absorbents
Leverandør: NEW PIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MAT304 PIG309
121-0052EA 1210 DKK
121-0052 NEWPPIG309
PIG® encapsulating and neutralizing absorbents
Spild materialer
One easy step for safer acid or caustic cleanup.
  • Choose from mats and socks to help encapsulate and neutralise common acids or caustics in one step
  • Colour changes as liquids are taken in, then remains in a colour-changed state when neutralisation is complete; helps identify the best time to handle with the least amount of risk
  • Instantly absorbs up to 98% concentrations of sulfuric acid
  • Gelling agents solidify acids during neutralisation; prevents them from leaching out of sock
Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.