Specialty solid phase extraction cartridges, Bond Elut™ Carbon/NH₂
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
446424 466430 12252202 12252201 12102201 126414 126418 2264265032
VARI446424EA 10530 DKK
VARI446424 VARI466430 VARI12252202 VARI12252201 VARI12102201 VARI126414 VARI126418 VARI2264265032
Specialty solid phase extraction cartridges, Bond Elut™ Carbon/NH₂
Kromatografikolonner
These specialty solid phase extraction cartridges provide higher retention of small organics, including those that are too polar to retain on C18 or polymeric SPE.
  • Typical matrices include organic plant and tissue extracts
  • Removal of chlorophyll and other pigments leads to fewer chromatographic or mass interferences
  • Broader retention and easier elution of analytes across the polarity range, for improved multi-residue analysis
These cartridges are packed with ultra-pure graphitised carbon particles that have been optimised for the absorption of pigments in food, fruits and vegetables, and small organic residues in waste water.