Flame-retardant overalls, ProShield® 20 SFR, model CHF5
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PFRCHF5SWH/XXL PFRCHF5SWH/3XL PFRCHF5SWH/XL PFRCHF5SWH00/L PFRCHF5SWH00/M
113-6384EA 4820 DKK
113-6384 113-8751 113-8752 113-8753 113-8754
Flame-retardant overalls, ProShield® 20 SFR, model CHF5
Engangsbeklædning
These hooded protective coveralls are made from a non halogenated, flame-retardant polypropylene SMS non woven fabric providing limited protection against flame spread (index 1) in addition to protection against particles and limited liquid splashes or sprays of water-based liquids. They must always be worn on top of a primary FR protection garment, such as Nomex®.
  • Zipper flap for protection
  • 3-piece hood and 3-piece gusset for optimal fit
  • Elasticated face, wrists, waist and ankles
  • Stitched external seams in orange for visual identification and differentiation
  • Generous fit offering high freedom of movement when wearing index 2 or 3 flame-retardant workwear beneath
  • Antistatic treatment (EN 1149-5) - on both sides
The overalls are used across a range of applications, including those in the petrochemical and railway industries, welding, gas and metal applications and certain Ex-Zones.
Colour: White (with orange seams)
Weight: 60 g/m²
Certificeringer: Type 5/6, EN 1149-5, EN 1073-2, EN ISO 14116-2008 (index 1); Cat. III.; (EU) 2016/425
Emballering: 50 per box, individually packed.