VWR®, Flasks, round bottom, with two necks and standard ground joints
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201-0542EA 206 DKK
201-0542 201-0540 201-0541 201-0543 201-0545 201-0544
VWR®, Flasks, round bottom, with two necks and standard ground joints
Kolber Kolber med rund bund
Borosilicate 3.3 glass, clear, with angled side neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
DIN 12394
Certificeringer: DIN 12394.