VWR®, Flasks, round bottom, with three necks and standard ground joints
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201-0546EA 186 DKK
201-0546 201-0547 201-0548 201-0549 201-0552
VWR®, Flasks, round bottom, with three necks and standard ground joints
Kolber Kolber med rund bund
Borosilicate 3.3 glass, clear, with two angled side necks.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
DIN 12394
Certificeringer: DIN 12394.