VWR®, Flasks, round bottom with three necks and standard ground joints
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201-0554EA 296 DKK
201-0554 201-0555 201-0553 201-0557 201-0556
VWR®, Flasks, round bottom with three necks and standard ground joints
Kolber Kolber med rund bund
Borosilicate 3.3 glass, clear, with two parallel side necks.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
An inexpensive alternative for a wide range of laboratory applications. With white labelling area and imprinted catalogue number for convenient reordering.
Certificeringer: DIN 12394