Mini desiccator cabinets, SICCO, Black
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
V1840-06 V1940-06
467-0124EA 2490 DKK
467-0124 467-0127
Mini desiccator cabinets, SICCO, Black
Ekssikatorer Ekssikatorkabinetter
Opaque black polycarbonate panels reduce light incidence and protect samples from ultraviolet radiation. Door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight, portable and stackable
  • Premium model includes hygrometer and two shelves
Leveringsinformation: Supplied with desiccant which is in a handy, resealable container.