TCLP testing filters, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5925-090 1810-110 1810-047 1810-090 1810-125 1810-142 1810-150 5925-142
WHAT5925-090EA 1140 DKK
WHAT5925-090 WHAT1810-110 WHAT1810-047 WHAT1810-090 WHAT1810-125 WHAT1810-142 WHAT1810-150 WHAT5925-142
TCLP testing filters, Whatman™
Filtre Glasfiberfiltre
Cytiva's Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Toxicity Characteristic Leaching Procedure (TCLP) is designed to predict the toxicity of hazardous waste. Industrial waste must comply with established TCLP limits to ensure that hazardous contaminants, known as landfill leachate, do not migrate into groundwater and threaten local drinking water sources.
By accurately characterizing the degree of contamination, material can be disposed of as safely and economically as possible.
Acid-treated, low metal Whatman TCLP test filters from Cytiva's business can help to determine what steps, if any, must be taken to mitigate landfill leachate risk.