Isoluminol (4-aminophthalhydrazide) 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

J64921.03 J64921.06
J64921.03EA 1290 DKK
J64921.03 J64921.06
Isoluminol (4-aminophthalhydrazide) 98%
Isoluminol (4-aminophthalhydrazide)
Formel: C₈H₇N₃O₂
Smeltepunkt: 300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010560
CAS-nummer: 3682-14-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR