Lithiumchlorid 1 M in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J61895.AE J61895.AD
J61895.AEEA 893 DKK
J61895.AE J61895.AD
Lithiumchlorid 1 M in aqueous solution
Lithium chloride
CAS No.: 7447-41-8 Molecular Formula: ClLi Formula Weight: 42.39 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00011078
Formel: LiCl
Molekylvægt: 42.39 g/mol
Kogepunkt: 1382
Smeltepunkt: 614
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011078
CAS-nummer: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550